EMAIL THIS PAGE TO A FRIEND
A2658 Sigma

Antipyrine

Sigma Reference Standard

Synonym: 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone

  • CAS Number 60-80-0

  • Empirical Formula (Hill Notation) C11H12N2O

  • Molecular Weight 188.23

  •  Beilstein Registry Number 157775

  •  EC Number 200-486-6

  •  MDL number MFCD00003146

  •  PubChem Substance ID 329770791

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   Sigma Reference Standard
InChI Key   VEQOALNAAJBPNY-UHFFFAOYSA-N
form   neat
packaging   vial of 250 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   109-111 °C(lit.)
format   neat

Description

Packaging

Supplied in amber screw-cap vials

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
CD2450000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A5882 Antipyrine, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?