• USA Home
  • A2681 - Anti-Aflatoxin B1−Peroxidase antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A2681 Sigma

Anti-Aflatoxin B1−Peroxidase antibody produced in rabbit

fractionated antiserum, buffered aqueous solution

Synonym: Anti-Aflatoxin B1

Purchase

Properties

Related Categories AE-AK, Alphabetical Index, Antibodies, Miscellaneous, Primary Antibodies More...
biological source   rabbit
antibody form   fractionated antiserum
clone   polyclonal
form   buffered aqueous solution
application(s)   direct ELISA: 1:1,000
conjugate   peroxidase conjugate
shipped in   dry ice
storage temp.   −20°C

Description

Immunogen

aflatoxin B1-KLH.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.01% thimerosal.

Application

Rabbit anti-aflatoxin B1-peroxidase antibody has been used for ELISA using peanut kernel homogenates.

General description

Aflatoxins are highly carcinogenic.

Aflatoxins are highly toxic mycotoxins produced by the fungus Aspergillus. Aflatoxins are commonly found in various grains and peanut butter.

Mycotoxins, a group of chemically diverse secondary fungal metabolites, induce a variety of toxic responses in humans and animals when food and feed containing these compounds are ingested. Rapidly quantitating levels of mycotoxin contamination can help reduce exposure to these toxins.

The antibody reacts with the B1 aflatoxin and also recognizes aflatoxins G1, B2, and the KLH protein. No cross-reaction is observed with aflatoxins and B2a, G2, G2a, M1.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?