• USA Home
  • A2735 - Anti-ANXA11 (111-125) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A2735 Sigma

Anti-ANXA11 (111-125) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-ANX11, Anti-Annexin A11, Anti-CAP50

Purchase

Properties

Related Categories AM-AN, Alphabetical Index, Antibodies, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~54 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ANXA11(311)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 111-125 of human ANXA11

General description

Annexin A11 (ANXA11) is a calcium-dependent phospholipid binding protein. Genetic polymorphisms of ANXA11 have been linked to the risk of sarcoidosis. Rabbit anti-ANXA11 (111-125) antibody binds to human ANXA11 (111-125).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Rabbit anti-ANXA11 (111-125) antibody can be used for western blot applications at dilutions ranging from 1:500-1:2,000.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?