• USA Home
  • A283 - 5′-Amino-5′-deoxyadenosine p-toluenesulfonate salt

EMAIL THIS PAGE TO A FRIEND
A283 Sigma

5′-Amino-5′-deoxyadenosine p-toluenesulfonate salt

solid

  • CAS Number 81090-75-7

  • Empirical Formula (Hill Notation) C10H14N6O3 · C7H8O3S

  • Molecular Weight 438.46

  •  MDL number MFCD00058132

  •  PubChem Substance ID 329770799

Purchase

Properties

InChI Key   KEAONEXDJAQJJU-MCDZGGTQSA-N
form   solid
color   white
solubility   H2O: soluble5 mg/mL
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?