• USA Home
  • A2889 - Albumin–fluorescein isothiocyanate conjugate

EMAIL THIS PAGE TO A FRIEND
A2889 Sigma

Albumin–fluorescein isothiocyanate conjugate

albumin from rat

Synonym: FITC-albumin

Purchase

Properties

biological source   albumin from rat
extent of labeling   ~10 mol FITC per mol albumin
storage temp.   2-8°C
Gene Information   rat ... Alb(24186)

Description

Preparation Note

Prepared from essentially globulin free albumin.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

A9771 Albumin–fluorescein isothiocyanate conjugate, albumin bovine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?