EMAIL THIS PAGE TO A FRIEND
A2915 Sigma

Propyl arachidonate

~90%

Synonym: cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid propyl ester, Arachidonic acid propyl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Esters, Fatty Acids and conjugates, Fatty Acyls, Lipids,
InChI Key   FVYDLXGAUHZBJE-GKFVBPDJSA-N
assay   ~90%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?