EMAIL THIS PAGE TO A FRIEND
A2935 Sigma

Aluminum oxide

Grade H-152, 1/8 in.

Synonym: Alumina

Purchase

Properties

InChI Key   TWNQGVIAIRXVLR-UHFFFAOYSA-N
type   Grade H-152
particle size   1/8 in.
mp   2040 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD1200000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?