EMAIL THIS PAGE TO A FRIEND
A3006 Sigma

Arabic acid

Purchase

Description

Linkage

This is a polysaccharide and should not be confused with L-arabinonic acid, the oxidation product of arabinose.

Packaging

5 g in poly bottle

Application

Arabic acid is a component of Gum Acacia.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?