• USA Home
  • A3019 - Adenosine 5′-monophosphate–Agarose

EMAIL THIS PAGE TO A FRIEND
A3019 Sigma

Adenosine 5′-monophosphate–Agarose

lyophilized powder, Contains lactose stabilizers that must be removed prior to use.

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Nucleotide/Coenzyme Resins, Protein Chromatography, Proteomics More...
form   lyophilized powder
extent of labeling   2-10 μmol per mL
matrix   cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   N-6
matrix spacer   8 atoms
storage temp.   −20°C

Description

Physical form

Lyophilized powder stabilized with lactose

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?