EMAIL THIS PAGE TO A FRIEND
A3039 Sigma

Wang resin

extent of labeling: 0.5-0.7 mmol per g resin

Synonym: p-Alkoxy-benzyl alcohol polymer-bound, p-Alkoxybenzyl alcohol resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene

Purchase

Properties

Related Categories Biochemicals and Reagents, Peptide Synthesis, Peptides More...
extent of labeling   0.5-0.7 mmol per g resin
storage temp.   2-8°C

Description

Application

For solid-phase peptide synthesis.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?