• USA Home
  • A3042 - Amyloglucosidase from Aspergillus niger

EMAIL THIS PAGE TO A FRIEND
A3042 Sigma

Amyloglucosidase from Aspergillus niger

aqueous glucose solution

Synonym: 1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Purchase

Description

Other Notes

View more information on enzymes for complex carbohydrate analysis at www.sigma-aldrich.com/enzymeexplorer

Unit Definition

One unit will liberate 1.0 mg of glucose from starch in 3 min at pH 4.5 at 55 °C.

Physical form

Solution in 1 M glucose containing 0.5% sodium benzoate as preservative

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A7095 Amyloglucosidase from Aspergillus niger, ≥260 U/mL, aqueous solution

inhibitor

Product #

Description

Add to Cart

D9305 1-Deoxynojirimycin hydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?