• USA Home
  • A3347 - Amyloid BRI Protein Precursor 227 Fragment 89-106

EMAIL THIS PAGE TO A FRIEND
A3347 Sigma

Amyloid BRI Protein Precursor 227 Fragment 89-106

≥81% (HPLC), solid

  • Empirical Formula (Hill Notation) C87H141N21O30S

  • Molecular Weight 1993.24

  •  MDL number MFCD03787948

Purchase

Description

Amino Acid Sequence

Cys-Gly-Ile-Lys-Tyr-Ile-Lys-Asp-Asp-Val-Ile-Leu-Asn-Glu-Pro-Ser-Ala-Asp

Biochem/physiol Actions

Peptide fragment found within the Amyloid BRI peptide.

Price and Availability


Amplified Detection

Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?