• USA Home
  • A3348 - ART® self-sealing barrier pipette tips
EMAIL THIS PAGE TO A FRIEND
A3348 Sigma
ART® self-sealing barrier pipette tips
ART 1000, volume range 100-1000 μL, sterile
Synonym: pipet tips, pipette tips
Purchase
Properties
Related Categories ART PCR Pipette Tips, ART self-sealing barrier pipette tips, Aerosol Barrier Pipette Tips, Labware, Liquid Handling Equipment,
material   colorless tip
sterility   sterile
feature   barrier
packaging   case of 4 packs (3200 tips)
  pkg of 800 tips
mfr. no.   Molecular BioProducts
volume range   100-1000 μL
Description
General description
New SoftFit design of the Art 1000 provides easy loading for a sure fit every time and easy ejecting for reduced hand fatigue.
Precision-molded tips with the Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling.
Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
Packaging
960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?