• USA Home
  • A3358 - AURORA C, active, GST tagged human

EMAIL THIS PAGE TO A FRIEND
A3358 Sigma

AURORA C, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: AIE2, AIK3, AURKC, AurC, STK13

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   55-75 nmol/min·mg
mol wt   mol wt ~61 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... AURKC(6795)

Description

Biochem/physiol Actions

Aurora C is a member of mitotic serine/threonine kinases that regulate centrosome maturation, chromosome segregation, and cytokinesis. Aurora C has been shown to contribute to oncogenic transformation and is frequently overexpressed and amplified in many human tumors. Aurora C overexpression has been correlated with centrosome amplification, which can be a driving cause of genomic instability in tumor cells. Aurora C is an important regulator of both genomic integrity and cell cycle progression in cancer cells and is an attractive target for anticancer drug development.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?