EMAIL THIS PAGE TO A FRIEND
A3434 Sigma

Albumin equine

~99% (agarose gel electrophoresis), lyophilized powder

Purchase

Properties

assay   ~99% (agarose gel electrophoresis)
form   lyophilized powder
impurities   globulin, essentially free
storage temp.   2-8°C

Description

Preparation Note

Prepared from albumin (A 9888).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?