• USA Home
  • A3483 - AURORA A, active, GST tagged from mouse

EMAIL THIS PAGE TO A FRIEND
A3483 Sigma

AURORA A, active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: AIK, ARK1, AURA, AURKA, BTAK, MGC34538, STK15, STK6, STK7

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   59-80 nmol/min·mg
mol wt   mol wt ~71 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   mouse ... Aurka(20878)

Description

Biochem/physiol Actions

AURORA A belongs to a multigenic family of mitotic serine/threonine kinases which are involved in the control of chromosome segregation. AURORA A is involved in centrosome separation, duplication and maturation as well as in bipolar spindle assembly and stability. AURORA A is expressed and active at the highest level during G2-M phase of the cell cycle. Overexpression of AURORA A has been found to be correlated with the grade of various human solid tumours. Ectopic AURORA A overexpression in any culture cell line leads to polyploidy and centrosome amplification.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?