• USA Home
  • A3521 - L-Alaninamide hydrobromide

EMAIL THIS PAGE TO A FRIEND
A3521 Sigma

L-Alaninamide hydrobromide

  • CAS Number 102029-80-1

  • Empirical Formula (Hill Notation) C3H8N2O · HBr

  • Molecular Weight 169.02

  •  MDL number MFCD00050707

Purchase

Properties

InChI Key   BYAVGTZKLGIZPY-DKWTVANSSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?