• USA Home
  • A3535 - α-Amylase Inhibitor from Triticum aestivum (wheat seed)

EMAIL THIS PAGE TO A FRIEND
A3535 Sigma

α-Amylase Inhibitor from Triticum aestivum (wheat seed)

Type III, lyophilized powder

Purchase

Properties

Related Categories A to C, Amylase, alpha-, Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Enzyme,
type   Type III
form   lyophilized powder
activity   3000-9000 inhibitor U/mg protein (using human salivary α-amylase)
  5-100 inhibitor U/mg protein (using porcine pancreatic α-amylase)
composition   Protein, 40-60% biuret
foreign activity   α-amylase ≤1 unit/mg protein
storage temp.   −20°C

Description

Unit Definition

One unit will reduce the activity of two units of α-amylase (A0521) by 50% after pre-incubation at 25 °C.

Physical form

Lyophilized powder containing buffer salts as sodium phosphate

Biochem/physiol Actions

Competitive inhibitor of human salivary α-amylase. KI = 2.9 nM, compared to a KM of 5.9 mM (calculated per mole of α-1,4-linked maltose residues).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?