• USA Home
  • A3640 - Adenosine 3′,5′-diphosphate–Agarose

EMAIL THIS PAGE TO A FRIEND
A3640 Sigma

Adenosine 3′,5′-diphosphate–Agarose

lyophilized powder

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Nucleotide/Coenzyme Resins, Protein Chromatography, Proteomics More...
form   lyophilized powder
extent of labeling   2-10 μmol per mL
matrix   cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   N-6
matrix spacer   8 atoms
storage temp.   −20°C

Description

Packaging

5 mL in poly bottle

Physical form

Lyophilized powder stabilized with lactose

Application

Adenosine 3′,5′-diphosphate Agarose (3′,5′-ADP agarose) has been used as an inexpensive medium to produce high yield Cytochrome P450 102, which is an important mammalian model of the P450 monooxygenase system.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?