• USA Home
  • A3668 - Anti-Angiotensin I antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A3668 Sigma

Anti-Angiotensin I antibody produced in rabbit

whole antiserum, lyophilized powder

Purchase

Properties

Related Categories AM-AN, Alphabetical Index, Antibodies, Antibodies for Neurotransmission, Antibodies to Neuropeptides,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
form   lyophilized powder
species reactivity   wide range
application(s)   radioimmunoassay: 1:10
conjugate   unconjugated
storage temp.   2-8°C

Description

Immunogen

angiotensin l-Asp1-lle5-BSA.

Reconstitution

Reconstitute with 5 mL buffer: 0.15 M phosphate buffer, pH 7.5, 0.03 M EDTA, 0.1% human serum albumin and 0.1% sodium azide. Store the reconstituted stock at −20 °C. Working dilutions to be discarded if unused within 12 hours.

Application

Rabbit anti-angiotensin I antibody can be used for radioimmunoassay at a dilution of 1:10.

General description

Angiotensin I is part of the renin-angiotensin system which regulates blood pressure by blood vessel constriction. Ang1 does not have biological activity but is a precursor to angiotensin II which is converted by ACE.

Rabbit anti-angiotensin I antibody does not cross-react with angiotensin II or angiotensin III by RIA. However, the antibody cross-reacts with Asn1-Val5-Angiotensin I.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?