• USA Home
  • A3793 - Anti-Aldosterone antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A3793 Sigma

Anti-Aldosterone antibody produced in rabbit

whole antiserum, lyophilized powder

Purchase

Properties

Related Categories AL-AL, Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Hormones,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
form   lyophilized powder
application(s)   radioimmunoassay: suitable
conjugate   unconjugated
storage temp.   2-8°C

Description

Immunogen

aldosterone-3-(O-carboxymethyloxime-BSA.

Physical form

The product is provided as a pre-diluted antiserum that has been lyophilized.

Application

Rabbit anti-aldosterone antibody can be used for radioimmunoassay.

General description

Aldosterone is a mineralocorticoid produced by the adrenal cortex. This hormone can induce urinary excretion of K+ and renal reabsorption of Na+. Rabbit Anti-Aldosterone antibody binds to aldosterone.

Price and Availability


Antibody Explorer
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Search 80,000 Antibodies at Sigma-Aldrich

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?