• USA Home
  • A3857 - Anti-AGK (241-254) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A3857 Sigma

Anti-AGK (241-254) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-Acylglycerol kinase, Anti-MULK, MAGK

Properties

Related Categories AE-AK, Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Lipids in Cell Signaling,
species reactivity   human
application(s)   indirect immunofluorescence: suitable
  western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~47 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... AGK(55750)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 241-254 of human AGK.

General description

AGK is a acylglycerol kinase that functions in glycerolipid metabolism by catalyzing the phosphorylation of monoacylglycerol.

Rabbit anti-AGK (241-254) antibody binds to human AGK (241-254).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.2, containing 15 mM sodium azide.

Application

Rabbit anti-AGK (241-254) antibody can be used for western blot (1:500-1:2000) and indirect immunofluorescence assays.

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?