EMAIL THIS PAGE TO A FRIEND
A3899 Sigma

Aminorex

Synonym: 4,5-Dihydro-5-phenyl-2-oxazolamine, Aminoxaphen

  • CAS Number 2207-50-3

  • Empirical Formula (Hill Notation) C9H10N2O

  • Molecular Weight 162.19

  •  MDL number MFCD00214070

Purchase

Properties

InChI Key   SYAKTDIEAPMBAL-UHFFFAOYSA-N
form   neat
drug control   USDEA Schedule I; Home Office Schedule 4.1; psychotrope; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
RQ4500000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?