EMAIL THIS PAGE TO A FRIEND
A3907 Sigma

Succinyl-Avidin

Synonym: Avidin-succinyl

Purchase

Properties

Related Categories Antibodies, Avidin Reagents, Biotin/Avidin, Supplementary Products
composition   Protein, ~90% E1%/280
storage temp.   −20°C

Description

Preparation Note

Prepared by succinylation by a modification of the procedure of Klapper, M.H. and Klotz, I.M., Meth. Enzymol., 25, 531 (1972).

Physical form

Lyophilized powder containing sodium citrate.

General description

Avidin is homotetrameric protein (68kDa) that is obtained from egg whites. This egg protein binds strongly to biotin. Thus, avidin-biotin association has been utilized in immunoassays to detect the localization of antigens in tissues. The use of avidin-biotin immunoassay enhances the sensitivity of the technique and facilitates the detection of antigens in low quantities.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?