• USA Home
  • A3909 - L-Arginine hydrochloride

EMAIL THIS PAGE TO A FRIEND
A3909 Sigma

L-Arginine hydrochloride

cell culture tested, insect cell culture tested

Purchase

Properties

suitability   cell culture tested
  insect cell culture tested

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
CF1995500

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A6969 L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?