• USA Home
  • A3927 - N-Acetyl-L-aspartic acid α-(p-nitroanilide)

EMAIL THIS PAGE TO A FRIEND
A3927 Sigma

N-Acetyl-L-aspartic acid α-(p-nitroanilide)

  • CAS Number 41149-01-3

  • Empirical Formula (Hill Notation) C12H13N3O6

  • Molecular Weight 295.25

  •  MDL number MFCD00078863

  •  PubChem Substance ID 24890734

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   CZAFZGHQNCYTDG-JTQLQIEISA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?