• USA Home
  • A4099 - Acetyl-γ-cyclodextrin

EMAIL THIS PAGE TO A FRIEND
A4099 Sigma

Acetyl-γ-cyclodextrin

  • Empirical Formula (Hill Notation) C96H120O64

  • Molecular Weight 2297.94

  •  MDL number MFCD03452813

  •  PubChem Substance ID 329770849

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Cyclodextrins, Synthetic Reagents
InChI Key   QNLDGLBAHVLKKB-BRHYBXGJSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?