• USA Home
  • A4147 - 3-Amino-1-propanesulfonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
A4147 Sigma

3-Amino-1-propanesulfonic acid sodium salt

≥98%

Purchase

Properties

Related Categories Agonists, Cell Biology, Cell Signaling and Neuroscience, GABA and Glycine Receptor Modulators, GABAergics,
InChI Key   YXTWMDHKIFWADD-UHFFFAOYSA-N
assay   ≥98%
storage temp.   2-8°C
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562)

Description

Biochem/physiol Actions

GABAA receptor agonist.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?