• USA Home
  • A4223 - [Phe10]-Amyloid β-Protein Fragment 1-40

EMAIL THIS PAGE TO A FRIEND
A4223 Sigma

[Phe10]-Amyloid β-Protein Fragment 1-40

≥95% (HPLC), lyophilized powder

Purchase

Properties

Related Categories Alzheimer′s Disease, Amyloid Precursor Proteins and Fragments, Amyloid beta Protein Fragments, Cell Biology, Cell Signaling and Neuroscience,
assay   ≥95% (HPLC)
form   lyophilized powder
mol wt   mol wt 4315
composition   Peptide content, ≥80%
solubility   H2O: soluble
storage temp.   −20°C
Gene Information   human ... APP(351)

Description

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Phe-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

Application

Used to study the effect of Phe10 on Aβ aggregation.

Price and Availability


Making Headway

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?