• USA Home
  • A4285 - N-(ε-Aminocaproyl)-β-L-fucopyranosylamine 4% beaded agarose

EMAIL THIS PAGE TO A FRIEND
A4285 Sigma

N-(ε-Aminocaproyl)-β-L-fucopyranosylamine 4% beaded agarose

buffered aqueous suspension

Purchase

Properties

form   buffered aqueous suspension
storage temp.   2-8°C

Description

Physical form

Suspension in 0.01 M phosphate buffer, pH 6.8, containing 0.15 M NaCl and 0.01% thimerosal

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?