• USA Home
  • A4473 - Amyloid β-Protein Fragment 1-40 amide trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
A4473 Sigma

Amyloid β-Protein Fragment 1-40 amide trifluoroacetate salt

≥95% (HPLC), lyophilized powder

  • Empirical Formula (Hill Notation) C194H296N54O57S · xC2HF3O2

  • Molecular Weight 4328.82 (free base basis)

  •  MDL number MFCD03454656

Purchase

Properties

Related Categories Alzheimer′s Disease, Amyloid Precursor Proteins and Fragments, Amyloid beta Protein Fragments, Cell Biology, Cell Signaling and Neuroscience,
assay   ≥95% (HPLC)
form   lyophilized powder
mol wt   mol wt 4331
composition   Protein content, ≥70%
solubility   H2O: soluble
storage temp.   −20°C
Gene Information   human ... APP(351)

Description

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-NH2

Other Notes

Amide form of Aβ [1-40]

Price and Availability


Making Headway

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?