EMAIL THIS PAGE TO A FRIEND
A4534 Sigma

L-Aspartic acid

BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym: (S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid

  • CAS Number 56-84-8

  • Linear Formula HO2CCH2CH(NH2)CO2H

  • Molecular Weight 133.10

  •  Beilstein Registry Number 1723530

  •  EC Number 200-291-6

  •  MDL number MFCD00002616

  •  PubChem Substance ID 24890873

  •  eCl@ss 32160406

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Amino Acids & Vitamins, Asymmetric Synthesis, Cell Culture, Chemical Synthesis, Chiral Resolution Reagents,
InChI Key   CKLJMWTZIZZHCS-REOHCLBHSA-N
product line   BioReagent
assay   ≥98% (TLC)
mp   >300 °C (dec.)(lit.)
suitability   suitable for cell culture
  suitable for insect cell culture
Gene Information   human ... CA1(759), CA2(760)
rat ... Grin2a(24409)

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
CI9098500
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?