• USA Home
  • A4594 - Anti-ADP Ribosylation Factor 1/3 (ARF 1/3) antibody produced in sheep

EMAIL THIS PAGE TO A FRIEND
A4594 Sigma

Anti-ADP Ribosylation Factor 1/3 (ARF 1/3) antibody produced in sheep

IgG fraction of antiserum, buffered aqueous solution

Properties

Related Categories AD-AD, Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to ARF and Related Proteins,
species reactivity   human, mouse
application(s)   western blot: 1:500-1:2,000 using RIPA lysates of mouse brain cells
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 21 kDa
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... ARF1(375), ARF3(377)
mouse ... Arf1(11840), Arf3(11842)
biological source   sheep
conjugate   unconjugated

Description

Immunogen

peptide corresponding to amino acids 98-112. Selected for maximal sequence homology with human ARF 1 and 3, but minimal sequence homology to other known ARFs.

Physical form

Solution in glycine buffered saline, containing 0.1% EACA, 0.01% benzamidine, 1 mM EDTA, and 0.1% sodium azide.

Preparation Note

The antibody is supplied as a DEAE purified IgG fraction.

Price and Availability


Detect Your Unseen Protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?