EMAIL THIS PAGE TO A FRIEND
A4640 Sigma
Amylopectin Azure
pectinase substrate
Purchase
Description
Other Notes
Soluble potato amylopectin covalently linked with Remazol Brilliant Blue R.
Substrates
Substrate for colorimetric determination of α-amylase.
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?