• USA Home
  • A4736 - AXL (473-end), active, His tagged human

EMAIL THIS PAGE TO A FRIEND
A4736 Sigma

AXL (473-end), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: JTK11, UFO

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   88-120 nmol/min·mg
mol wt   mol wt ~55 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... AXL(558)

Description

Biochem/physiol Actions

AXL is a member of the receptor tyrosine kinase family which has oncogenic potential and is implicated in human myeloid leukemia. AXL is a member of a complex signaling network that is involved in the control of cell proliferation and differentiation. Overexpression of AXL cDNA in NIH 3T3 cells induces neoplastic transformation of these cells with the concomitant appearance of a 140 kDa AXL tyrosine-phosphorylated protein. Expression of AXL cDNA in the baculovirus system results in the expression of the appropriate recombinant protein that is recognized by antiphosphotyrosine antibodies, confirming that the AXL protein is tyrosine phosphorylated.

Physical form

Supplied in 50 mM Sodium Phosphate, pH 7.0, with 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?