• USA Home
  • A4754 - 4-Amino-1,3-dimethyl-2,6-dioxy-5-nitrosopyrimidine

EMAIL THIS PAGE TO A FRIEND
A4754 Sigma

4-Amino-1,3-dimethyl-2,6-dioxy-5-nitrosopyrimidine

Synonym: 6-Amino-1,3-dimethyl-5-nitrosouracil

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   MGBDANYXBKROBW-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YQ8780000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?