• USA Home
  • A4887 - Asparaginase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
A4887 Sigma

Asparaginase from Escherichia coli

Grade V, buffered aqueous glycerol solution, ≥80 units/mg protein

Synonym: L-Asparagine Amidohydrolase

Purchase

Description

Analysis Note

Protein determined by biuret.

Unit Definition

One unit will liberate 1.0 μmole of ammonia from L-asparagine per min at pH 8.6 at 37 °C.

Physical form

Solution in 50% glycerol, pH 6.5

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A3809 Asparaginase from Escherichia coli, lyophilized powder, 100-300 units/mg protein (biuret)

inhibitor

Product #

Description

Add to Cart

C5913 4-Chloromercuribenzoic acid, cysteine active site modifier

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?