• USA Home
  • A4893 - ω-Aminopropyl–Agarose

EMAIL THIS PAGE TO A FRIEND
A4893 Sigma

ω-Aminopropyl–Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Hydrophobic Interaction, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
extent of labeling   3-5 μmol per mL
matrix   4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino of 1,3-diaminopropane
matrix spacer   1 atom
capacity   2-4 mg/mL binding capacity (bovine serum albumin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?