• USA Home
  • A4930 - N-Acetyl-D-galactosaminitol

EMAIL THIS PAGE TO A FRIEND
A4930 Sigma

N-Acetyl-D-galactosaminitol

Synonym: 2-Acetamido-2-deoxy-D-galactitol

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Monosaccharide More...
InChI Key   DWAICOVNOFPYLS-OSMVPFSASA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?