EMAIL THIS PAGE TO A FRIEND
A5020 Sigma

Acid red 183

  •  Colour Index Number 18800

  •  EC Number 229-054-5

Purchase

Properties

composition   Dye content, ~60%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?