• USA Home
  • A5032 - Adenosine 5′-diphosphate lithium salt

EMAIL THIS PAGE TO A FRIEND
A5032 Sigma

Adenosine 5′-diphosphate lithium salt

recombinant, expressed in bacteria, 95-99%

Synonym: ADP

  • CAS Number 31008-64-7

  • Empirical Formula (Hill Notation) C10H15N5O10P2

  • Molecular Weight 427.20

  •  EC Number 250-431-5

Purchase

Properties

recombinant   expressed in bacteria
assay   95-99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

A2754 Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?