• USA Home
  • A5324 - Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in sheep

EMAIL THIS PAGE TO A FRIEND
A5324 Sigma

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in sheep

affinity isolated antibody, buffered aqueous glycerol solution

Purchase

Properties

Related Categories Alkaline Phosphatase, Alphabetical Index, Antibodies, MO-MR, Mouse Secondary Antibodies and Conjugates,
biological source   sheep
antibody form   affinity isolated antibody
clone   polyclonal
form   buffered aqueous glycerol solution
application(s)   direct ELISA: 1:30,000
  immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
  western blot: 1:30,000
conjugate   alkaline phosphatase conjugate
shipped in   wet ice
storage temp.   2-8°C

Description

Other Notes

Antibody adsorbed with human serum proteins.

Preparation Note

Adsorbed to reduce background with human samples.

Specificity

Binds all mouse Igs.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 50% glycerol, 10 mM glycine, and 15 mM sodium azide.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?