• USA Home
  • A5338 - p-Amino-D-phenylalanine hydrochloride

EMAIL THIS PAGE TO A FRIEND
A5338 Sigma

p-Amino-D-phenylalanine hydrochloride

  • CAS Number 196408-63-6

  • Empirical Formula (Hill Notation) C9H12N2O2 · HCl

  • Molecular Weight 216.66

  •  MDL number MFCD00236783

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   ISCZSPZLBJLJGO-DDWIOCJRSA-N
assay   ≥99% (TLC)
form   white powder
solubility   H2O: soluble20 mg/mL
  1 M HCl: soluble50 mg/mL
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?