• USA Home
  • A5345 - N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin

EMAIL THIS PAGE TO A FRIEND
A5345 Sigma

N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin

≥97% (HPLC), powder

Synonym: Ac-VDVAD-AFC

Purchase

Description

Amino Acid Sequence

NAc-Val-Asp-Val-Ala-Asp-AFC

Substrates

Fluorogenic substrate for caspase 2.

Price and Availability


Amplified Detection

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?