• USA Home
  • A5357 - Anti-ALK (1601-1615) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A5357 Sigma

Anti-ALK (1601-1615) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-Anaplastic lymphoma kinase Ki-1, Anti-CD246, Anti-TFG/ALK

Purchase

Properties

Related Categories AL-AL, ALK, Alphabetical Index, Antibodies, Antibodies for Cell Biology,
species reactivity   human
application(s)   western blot: 1:1,000-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~176 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ALK(238)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 1601-1615 of human ALK.

General description

Anaplastic lymphoma receptor tyrosine kinase (ALK) is a tyrosine kinase member of the superfamily of insulin receptors. ALK modulates neurological development. Genetic alterations in ALK have been linked to neurological, lung and white blood cell cancers. Rabbit Anti- ALK (1601-1615) binds to human ALK (1601-1615).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.2, containing 15 mM sodium azide.

Application

Rabbit Anti-ALK (1601-1615) antibody can be used for western blot (1:1,000-1:2,000) assays.

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

HPA010694 Anti-ALK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?