• USA Home
  • A5381 - L-Arginine Decarboxylase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
A5381 Sigma

L-Arginine Decarboxylase from Escherichia coli

Type II, lyophilized powder, 5-15 units/mg protein

Synonym: L-Arginine carboxylyase, ADC, Arginine decarboxylase

Purchase

Properties

Related Categories 4.1.x.x C-C bonds, 4.x.x.x Lyases, Amino Acid Metabolism, Biochemicals and Reagents, Cell Biology,
biological source   from ATCC 10787
type   Type II
form   lyophilized powder
composition   Protein, ~50% biuret
storage temp.   −20°C

Description

Unit Definition

One unit will release 1.0 μmole of CO2 from L-arginine per min at pH 5.2 at 37 °C.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?