• USA Home
  • A5411 - 4-Amino-3-nitrotoluene

EMAIL THIS PAGE TO A FRIEND
A5411 Sigma

4-Amino-3-nitrotoluene

~99%

Synonym: 2-Nitro-p-toluidine, 4-Methyl-2-nitroaniline, Fast red gl

  •  Colour Index Number 37110

  •  EC Number 201-924-9

Purchase

Properties

assay   ~99%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2660 6.1 / PGIII
WGK Germany 
3
RTECS 
XU8227250

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?