EMAIL THIS PAGE TO A FRIEND
A5510 Sigma

N-Acetyl-Ala-Ala-Ala

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Peptides, Tripeptides
InChI Key   DRYOODAJROGPQO-ACZMJKKPSA-N
storage temp.   −20°C

Description

Amino Acid Sequence

NAc-Ala-Ala-Ala

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?