EMAIL THIS PAGE TO A FRIEND
A5524 Sigma

Acid Blue 45

  •  Colour Index Number 63010

  •  EC Number 220-678-3

Purchase

Properties

composition   Dye content, ~40%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
CB0548500

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?