EMAIL THIS PAGE TO A FRIEND
A5534 Sigma

N4-Anisoylcytidine

  • CAS Number 28225-17-4

  • Empirical Formula (Hill Notation) C17H19N3O7

  • Molecular Weight 377.35

  •  MDL number MFCD00057436

  •  PubChem Substance ID 24890989

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   UAHIASPEJUIQKN-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?